Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116461.4 | NP_192137.1 | 8 - 9 | 2129 | 81 | |
GenBank mRNA | AY051001.1 | - | 8 - 9 | 1903 | 81 | |
AY056784.1 | - | 8 - 9 | 1928 | 81 | ||
AY142511.1 | - | 8 - 9 | 1689 | 81 | ||
KJ138769.1 | - | 8 - 9 | 1689 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |