Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203731.2 | NP_001190660.1 | 11 - 12 | 1275 | 79 | |
NM_116490.4 | NP_192165.1 | 10 - 11 | 1418 | 79 | ||
GenBank mRNA | AK226746.1 | - | 10 - 11 | 1328 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |