Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203731.2 | NP_001190660.1 | 6 - 7 | 878 | 77 | |
NM_116490.4 | NP_192165.1 | 5 - 6 | 1021 | 77 | ||
GenBank mRNA | AK226746.1 | - | 5 - 6 | 931 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |