Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036498.3 | NP_001031575.1 | 16 - 17 | 1663 | 73 | |
NM_001036499.1 | NP_001031576.1 | 16 - 17 | 1599 | 73 | ||
NM_001203732.1 | NP_001190661.1 | 16 - 17 | 1661 | 73 | ||
NM_116491.3 | NP_567243.1 | 15 - 16 | 1757 | 73 | ||
GenBank mRNA | AJ318017.1 | - | 14 - 15 | 1458 | 73 | |
AK317123.1 | - | 16 - 17 | 1600 | 73 | ||
AY046030.1 | - | 15 - 16 | 1680 | 73 | ||
AY133878.1 | - | 14 - 15 | 1424 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |