Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZO2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340389.1 | NP_001329302.1 | 4 - 5 | 841 | 99 | |
NM_001340390.1 | NP_001329303.1 | 4 - 5 | 823 | 99 | ||
NM_116498.4 | NP_192173.1 | 4 - 5 | 823 | 99 | ||
NM_178959.3 | NP_849290.1 | 4 - 5 | 832 | 99 | ||
GenBank mRNA | AF310222.1 | - | 4 - 5 | 779 | 99 | |
AJ010859.1 | - | 4 - 5 | 771 | 99 | ||
AK318728.2 | - | 4 - 5 | 764 | 99 | ||
AY088417.1 | - | 4 - 5 | 754 | 99 | ||
BT025558.1 | - | 4 - 5 | 755 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |