Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116502.4 | NP_192177.1 | 2 - 3 | 2531 | 67 | |
GenBank mRNA | AK229195.1 | - | 2 - 3 | 2449 | 67 | |
AY572792.1 | - | 2 - 3 | 1847 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |