Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116512.2 | NP_192187.1 | 6 - 7 | 1009 | 62 | |
GenBank mRNA | U11034.1 | - | 6 - 7 | 1000 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |