Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116532.3 | NP_192207.1 | 2 - 3 | 386 | 67 | |
GenBank mRNA | AF389278.1 | - | 2 - 3 | 236 | 67 | |
AY093754.1 | - | 2 - 3 | 227 | 67 | ||
S40550.1 | - | 2 - 3 | 236 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |