Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD6;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340443.1 | NP_001329275.1 | 4 - 5 | 578 | 76 | |
NM_116565.2 | NP_192236.1 | 4 - 5 | 751 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |