Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD6;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340443.1 | NP_001329275.1 | 5 - 6 | 718 | 142 | |
NM_116565.2 | NP_192236.1 | 5 - 6 | 891 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |