Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL05 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116593.4 | NP_192264.1 | 4 - 5 | 5654 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |