Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116594.5 | NP_567258.1 | 5 - 6 | 921 | 80 | |
GenBank mRNA | AF360372.1 | - | 5 - 6 | 597 | 80 | |
AK316725.1 | - | 5 - 6 | 638 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |