Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340496.1 | NP_001319866.1 | 7 - 8 | 1406 | 111 | |
NM_001340497.1 | NP_001329214.1 | 7 - 8 | 1469 | 111 | ||
NM_001340498.1 | NP_001329213.1 | 8 - 9 | 1325 | 111 | ||
NM_001340499.1 | NP_001329212.1 | 7 - 8 | 1412 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |