Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203746.1 | NP_001190675.1 | 15 - 16 | 2212 | 108 | |
NM_116723.5 | NP_567272.4 | 15 - 16 | 1993 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |