Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116725.5 | NP_567273.1 | 4 - 5 | 824 | 65 | |
GenBank mRNA | AY059735.1 | - | 4 - 5 | 758 | 65 | |
AY091269.1 | - | 4 - 5 | 678 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |