Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116725.5 | NP_567273.1 | 6 - 7 | 987 | 135 | |
GenBank mRNA | AY059735.1 | - | 6 - 7 | 921 | 135 | |
AY091269.1 | - | 6 - 7 | 841 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |