Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340548.1 | NP_001328043.1 | 6 - 7 | 622 | 136 | |
NM_001340549.1 | NP_001328044.1 | 6 - 7 | 653 | 136 | ||
NM_116772.4 | NP_567292.1 | 6 - 7 | 699 | 136 | ||
GenBank mRNA | AY084733.1 | - | 6 - 7 | 569 | 136 | |
BT024733.1 | - | 5 - 6 | 510 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |