Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116790.3 | NP_567299.2 | 12 - 13 | 1221 | 99 | |
NM_202788.1 | NP_974517.1 | 12 - 13 | 1170 | 99 | ||
GenBank mRNA | BT033036.1 | - | 12 - 13 | 1102 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |