Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340568.1 | NP_001328773.1 | 4 - 5 | 1697 | 133 | |
NM_116866.3 | NP_192536.1 | 5 - 6 | 1709 | 133 | ||
GenBank mRNA | AK118480.1 | - | 5 - 6 | 1755 | 133 | |
AY087066.1 | - | 4 - 5 | 1565 | 133 | ||
BT008770.1 | - | 4 - 5 | 1474 | 133 | ||
KJ138765.1 | - | 4 - 5 | 1474 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |