Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116873.2 | NP_567330.1 | 2 - 3 | 426 | 108 | |
GenBank mRNA | AF332405.1 | - | 2 - 3 | 276 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |