Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116884.4 | NP_192555.1 | 1 - 2 | 1022 | 80 | |
GenBank mRNA | AY080834.1 | - | 1 - 2 | 578 | 80 | |
AY113982.1 | - | 1 - 2 | 473 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |