Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340578.1 | NP_001319881.1 | 8 - 9 | 2432 | 111 | |
NM_116887.3 | NP_192558.2 | 8 - 9 | 2407 | 111 | ||
NM_179012.2 | NP_849343.2 | 8 - 9 | 2404 | 111 | ||
NM_202789.1 | NP_974518.1 | 8 - 9 | 2404 | 111 | ||
GenBank mRNA | AY139750.1 | - | 8 - 9 | 2405 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |