Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340588.1 | NP_001329232.1 | 1 - 2 | 487 | 97 | |
NM_001340589.1 | NP_001329233.1 | 1 - 2 | 487 | 97 | ||
NM_116916.4 | NP_192587.1 | 1 - 2 | 689 | 97 | ||
GenBank mRNA | BT021102.1 | - | 1 - 2 | 487 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |