Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340588.1 | NP_001329232.1 | 2 - 3 | 740 | 67 | |
NM_116916.4 | NP_192587.1 | 2 - 3 | 942 | 67 | ||
GenBank mRNA | BT021102.1 | - | 2 - 3 | 740 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |