Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116931.3 | NP_192602.1 | 10 - 11 | 1716 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons |