Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC75-IV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340610.1 | NP_001329162.1 | 2 - 3 | 317 | 96 | |
NM_001340611.1 | NP_001329163.1 | 1 - 2 | 163 | 96 | ||
NM_116977.3 | NP_192647.2 | 3 - 4 | 409 | 96 | ||
GenBank mRNA | AY585655.1 | - | 3 - 4 | 386 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |