Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203413.1 | NP_001190342.1 | 6 - 7 | 830 | 76 | |
NM_121998.5 | NP_197491.2 | 5 - 6 | 658 | 76 | ||
GenBank mRNA | AY072222.1 | - | 5 - 6 | 610 | 76 | |
BT002744.1 | - | 5 - 6 | 571 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |