Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104102.3 | NP_175633.1 | 1 - 2 | 233 | 111 | |
GenBank mRNA | AF385712.1 | - | 1 - 2 | 183 | 111 | |
AJ245632.1 | - | 1 - 2 | 149 | 111 | ||
AY085302.1 | - | 1 - 2 | 195 | 111 | ||
AY133667.1 | - | 1 - 2 | 129 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |