Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036101.2 | NP_001031178.1 | 3 - 4 | 683 | 82 | |
NM_001036102.3 | NP_001031179.1 | 2 - 3 | 814 | 82 | ||
NM_104209.3 | NP_175738.1 | 3 - 4 | 744 | 82 | ||
GenBank mRNA | AJ532901.1 | - | 2 - 3 | 556 | 82 | |
AY057507.1 | - | 3 - 4 | 622 | 82 | ||
BT000647.1 | - | 2 - 3 | 556 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |