Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036101.2 | NP_001031178.1 | 5 - 6 | 1001 | 106 | |
NM_001036102.3 | NP_001031179.1 | 4 - 5 | 1132 | 106 | ||
NM_104209.3 | NP_175738.1 | 5 - 6 | 1062 | 106 | ||
GenBank mRNA | AJ532901.1 | - | 4 - 5 | 874 | 106 | |
AY057507.1 | - | 5 - 6 | 940 | 106 | ||
BT000647.1 | - | 4 - 5 | 874 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |