Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104236.5 | NP_564636.2 | 6 - 7 | 916 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |