Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104246.2 | NP_175772.1 | 1 - 2 | 358 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |