Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333607.1 | NP_001321367.1 | 4 - 5 | 439 | 106 | |
NM_104250.5 | NP_175776.2 | 4 - 5 | 446 | 106 | ||
GenBank mRNA | AY568526.1 | - | 3 - 4 | 320 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |