Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333607.1 | NP_001321367.1 | 9 - 10 | 989 | 69 | |
NM_104250.5 | NP_175776.2 | 9 - 10 | 996 | 69 | ||
GenBank mRNA | AY568526.1 | - | 8 - 9 | 870 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |