Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 4 - 5 | 589 | 78 | |
NM_179476.3 | NP_849807.1 | 3 - 4 | 475 | 78 | ||
GenBank mRNA | AF378873.1 | - | 4 - 5 | 447 | 78 | |
AJ584645.1 | - | 3 - 4 | 357 | 78 | ||
AK230363.1 | - | 2254 | 78 | |||
AY048242.1 | - | 3 - 4 | 472 | 78 | ||
AY091032.1 | - | 3 - 4 | 474 | 78 | ||
AY102145.1 | - | 3 - 4 | 355 | 78 | ||
AY117345.1 | - | 3 - 4 | 355 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |