Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 7 - 8 | 753 | 85 | |
NM_179476.3 | NP_849807.1 | 6 - 7 | 639 | 85 | ||
GenBank mRNA | AF378873.1 | - | 7 - 8 | 611 | 85 | |
AJ584645.1 | - | 6 - 7 | 521 | 85 | ||
AK230363.1 | - | 2418 | 85 | |||
AY048242.1 | - | 6 - 7 | 636 | 85 | ||
AY091032.1 | - | 6 - 7 | 638 | 85 | ||
AY102145.1 | - | 6 - 7 | 519 | 85 | ||
AY117345.1 | - | 6 - 7 | 519 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |