Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104295.4 | NP_175819.1 | 12 - 13 | 1879 | 62 | |
GenBank mRNA | AY136398.1 | - | 12 - 13 | 1770 | 62 | |
BT000204.1 | - | 11 - 12 | 1562 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |