Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104295.4 | NP_175819.1 | 9 - 10 | 1490 | 83 | |
GenBank mRNA | AY136398.1 | - | 9 - 10 | 1381 | 83 | |
BT000204.1 | - | 8 - 9 | 1173 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |