Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333641.1 | NP_001321996.1 | 9 - 10 | 800 | 111 | |
NM_104312.3 | NP_175836.1 | 9 - 10 | 867 | 111 | ||
GenBank mRNA | AF316501.1 | - | 9 - 10 | 693 | 111 | |
BT025983.1 | - | 9 - 10 | 673 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |