Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198295.1 | NP_001185224.1 | 2 - 3 | 192 | 72 | |
NM_001198296.1 | NP_001185225.1 | 2 - 3 | 127 | 72 | ||
NM_001333653.1 | NP_001319226.1 | 1 - 2 | 535 | 72 | ||
NM_104329.5 | NP_175853.1 | 2 - 3 | 288 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |