Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333664.1 | NP_001323099.1 | 4 - 5 | 1915 | 76 | |
NM_104346.3 | NP_564664.1 | 5 - 6 | 2496 | 76 | ||
GenBank mRNA | AY142626.1 | - | 5 - 6 | 2073 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |