Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRUCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124022.2 | NP_001117494.1 | 4 - 5 | 1563 | 153 | |
NM_001198301.1 | NP_001185230.1 | 4 - 5 | 1442 | 153 | ||
NM_001333690.1 | NP_001320753.1 | 4 - 5 | 1739 | 153 | ||
NM_104385.4 | NP_564676.1 | 5 - 6 | 1583 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |