Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160956.1 | NP_001154428.1 | 4 - 5 | 473 | 107 | |
NM_001198304.1 | NP_001185233.1 | 15 - 16 | 2116 | 107 | ||
NM_001198305.2 | NP_001185234.1 | 4 - 5 | 917 | 107 | ||
NM_001333696.1 | NP_001321104.1 | 14 - 15 | 2181 | 107 | ||
NM_104398.4 | NP_564680.4 | 14 - 15 | 1977 | 107 | ||
GenBank mRNA | BT029207.1 | - | 4 - 5 | 426 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |