Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 11 - 12 | 1776 | 75 | |
NM_104408.5 | NP_175929.3 | 11 - 12 | 2039 | 75 | ||
GenBank mRNA | AY250845.1 | - | 11 - 12 | 1764 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |