Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPN60B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333710.1 | NP_001320639.1 | 11 - 12 | 1924 | 116 | |
NM_001333711.1 | NP_001320638.1 | 11 - 12 | 1907 | 116 | ||
NM_001333712.1 | NP_001320637.1 | 11 - 12 | 1890 | 116 | ||
NM_104424.2 | NP_175945.1 | 11 - 12 | 1615 | 116 | ||
NM_179480.3 | NP_849811.1 | 11 - 12 | 1686 | 116 | ||
GenBank mRNA | AF386945.1 | - | 11 - 12 | 1607 | 116 | |
AK316889.1 | - | 11 - 12 | 1592 | 116 | ||
AY081501.1 | - | 10 - 11 | 1485 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |