Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198311.2 | NP_001185240.1 | 6 - 7 | 1676 | 64 | |
NM_001198312.2 | NP_001185241.1 | 6 - 7 | 1637 | 64 | ||
NM_001333713.1 | NP_001321787.1 | 6 - 7 | 1676 | 64 | ||
NM_001333714.1 | NP_001321789.1 | 6 - 7 | 1676 | 64 | ||
NM_001333715.1 | NP_001321788.1 | 5 - 6 | 1851 | 64 | ||
NM_104425.1 | NP_564692.1 | 5 - 6 | 1785 | 64 | ||
GenBank mRNA | AY050425.1 | - | 5 - 6 | 1785 | 64 | |
BT002240.1 | - | 4 - 5 | 1234 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |