Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104433.5 | NP_175953.1 | 2 - 3 | 1483 | 66 | |
GenBank mRNA | AF367325.1 | - | 2 - 3 | 1483 | 66 | |
AY133602.1 | - | 2 - 3 | 1368 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |