Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104438.4 | NP_564698.1 | 5 - 6 | 1829 | 86 | |
NM_179482.3 | NP_849813.1 | 5 - 6 | 1829 | 86 | ||
GenBank mRNA | AF366368.1 | - | 5 - 6 | 1691 | 86 | |
AJ303348.1 | - | 5 - 6 | 1695 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |