Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104450.4 | NP_175970.1 | 9 - 10 | 1604 | 85 | |
GenBank mRNA | AK227977.1 | - | 9 - 10 | 1626 | 85 | |
BT004640.1 | - | 9 - 10 | 1533 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |