Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104462.4 | NP_175981.2 | 2 - 3 | 1191 | 96 | |
GenBank mRNA | AK227116.1 | - | 2 - 3 | 933 | 96 | |
AY064040.1 | - | 2 - 3 | 932 | 96 | ||
KJ138961.1 | - | 2 - 3 | 879 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |